tert-Butyl-4-[4-methyl-5-(methylsulphanyl)-4H-1,2,4-triazol-3-yl]tetrahydro-1(2H)-pyridine carboxylate
Leverantör: Apollo Scientific
Synonyms: 4-[4-Methyl-5-(methylthio)-4H-1,2,4-triazol-3-yl]piperidine, N-BOC protected
OR8516-1G OR8516-5G OR8516-10G
APOSOR8516-1GEA 3400 SEK
APOSOR8516-1G APOSOR8516-5G APOSOR8516-10G
tert-Butyl-4-[4-methyl-5-(methylsulphanyl)-4H-1,2,4-triazol-3-yl]tetrahydro-1(2H)-pyridine carboxylate
tert-Butyl-4-[4-methyl-5-(methylsulphanyl)-4H-1,2,4-triazol-3-yl]tetrahydro-1(2H)-pyridine carboxylate
Formel: C₁₄H₂₄N₄O₂S
MW: 312,44 g/mol
Smältpunkt: 116…118 °C
Storage Temperature: Omgivande
MDL Number: MFCD09607934
CAS nummer: 952183-43-6