1-(4-Chlorobenzyl)-1H-1,2,3-triazole-4-carbaldehyde

Leverantör: Apollo Scientific

OR8497-1G OR8497-500MG
APOSOR8497-1GEA 5125 SEK
APOSOR8497-1G APOSOR8497-500MG
1-(4-Chlorobenzyl)-1H-1,2,3-triazole-4-carbaldehyde
1-(4-Chlorobenzyl)-1H-1,2,3-triazole-4-carbaldehyde
Formel: C₁₀H₈ClN₃O
MW: 221,65 g/mol
Smältpunkt: 92…94 °C
Storage Temperature: Omgivande
MDL Number: MFCD09607915
CAS nummer: 952183-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR