2-Bromobenzaldehyde diethyl acetal
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Bromo-2-(diethoxymethyl)benzene, 2-Bromo-α,α-diethoxytoluene
OR8458-100G OR8458-25G
APOSOR8458-100GEA 1100 SEK
APOSOR8458-100G APOSOR8458-25G
2-Bromobenzaldehyde diethyl acetal
2-Bromobenzaldehyde diethyl acetal
Formel: C₁₁H₁₅BrO₂
Kokpunkt: 95 °C/1mmHg
Densitet: 1,285 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD01075696
CAS nummer: 35822-58-3