2-Bromo-4'-(1-piperidinyl)acetophenone 98%
Leverantör: Apollo Scientific
Fara
Synonyms: 4-(Piperidin-1-yl)phenacyl bromide
OR8309-1G OR8309-5G
APOSOR8309-1GEA 1200 SEK
APOSOR8309-1G APOSOR8309-5G
2-Bromo-4'-(1-piperidinyl)acetophenone 98%
2-Bromo-4'-(1-piperidinyl)acetophenone
Formel: C₁₃H₁₆BrNO
Storage Temperature: Omgivande
MDL Number: MFCD06808683
CAS nummer: 210832-84-1
UN: 3261
ADR: 8,III