4-Nitrosophenol 98%
Leverantör: Apollo Scientific
Synonyms:
1,4-Benzoquinone Monoxime, p-Quinone Monoxime
OR8238-25G
OR8238-50G
OR8238-100G
APOSOR8238-25GEA
6000
SEK
APOSOR8238-25G
APOSOR8238-50G
APOSOR8238-100G
4-Nitrosophenol 98%
4-Nitrosophenol
Formel:
C₆H₅NO₂ MW: 123,11 g/mol Smältpunkt: 126…128 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00002329 CAS nummer: 104-91-6 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...