3,4-Diaminobenzenesulphonamide 95+%
Leverantör: Apollo Scientific
|
Varning
|
OR8069-1G
OR8069-250MG
OR8069-500MG
OR8069-2.5G
OR8069-5G
APOSOR8069-1GEA
3225
SEK
APOSOR8069-1G
APOSOR8069-250MG
APOSOR8069-500MG
APOSOR8069-2.5G
APOSOR8069-5G
3,4-Diaminobenzenesulphonamide 95+%
3,4-Diaminobenzenesulphonamide
Formel:
C₆H₉N₃O₂S Smältpunkt: 160 °C (dec.) Storage Temperature: Omgivande |
MDL Number:
MFCD00277940 CAS nummer: 2360-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...