1-Boc-4-piperidone 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone
OR8050-100G
OR8050-25G
APOSOR8050-100GEA
585
SEK
APOSOR8050-100G
APOSOR8050-25G
1-Boc-4-piperidone 98%
1-Boc-4-piperidone
Formel:
C₁₀H₁₇NO₃ MW: 199,25 g/mol Smältpunkt: 72…77 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00151800 CAS nummer: 79099-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...