5-(Benzyloxy)-1H-indole-2-carboxylic acid, Technical Grade
Leverantör: Apollo Scientific
Varning
OR8026-1G OR8026-500MG OR8026-5G OR8026-10G OR8026-25G
APOSOR8026-1GEA 1125 SEK
APOSOR8026-1G APOSOR8026-500MG APOSOR8026-5G APOSOR8026-10G APOSOR8026-25G
5-(Benzyloxy)-1H-indole-2-carboxylic acid, Technical Grade
5-(Benzyloxy)-1H-indole-2-carboxylic acid
Formel: C₁₆H₁₃NO₃
Smältpunkt: 194…200 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00047165
CAS nummer: 6640-09-1
EINECS: 229-652-6