1-[(4-Methylphenyl)sulphonyl]-1H-indol-5-amine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Amino-1-[(4-methylphenyl)sulphonyl]-1H-indole hydrochloride
OR8010-1G OR8010-5G OR8010-10G
APOSOR8010-1GEA 2150 SEK
APOSOR8010-1G APOSOR8010-5G APOSOR8010-10G
1-[(4-Methylphenyl)sulphonyl]-1H-indol-5-amine hydrochloride
1-[(4-Methylphenyl)sulphonyl]-1H-indol-5-amine hydrochloride
Formel: C₁₅H₁₅ClN₂O₂S
MW: 322,82 g/mol
Smältpunkt: 242…244 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02259420
CAS nummer: 306937-24-6