(+)-5,6-O-Isopropylidene-L-ascorbic acid
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-5-((S)-2,2-Dimethyl-1,3-dioxolan-4-yl)-3,4-dihydroxyfuran-2(5H)-one
OR7870T-25G OR7870T-5G
APOSOR7870T-25GEA 5675 SEK
APOSOR7870T-25G APOSOR7870T-5G
(+)-5,6-O-Isopropylidene-L-ascorbic acid
(+)-5,6-O-Isopropylidene-L-ascorbic acid
Formel: C₉H₁₂O₆
MW: 216,19 g/mol
Smältpunkt: 210 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00010552
CAS nummer: 15042-01-0