Cyclopentylboronic acid
Leverantör: Apollo Scientific
Varning
OR7833-1G OR7833-5G
APOSOR7833-1GEA 260 SEK
APOSOR7833-1G APOSOR7833-5G
Cyclopentylboronic acid
Cyclopentylboronic acid
Formel: C₅H₁₁BO₂
Smältpunkt: 126 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD01074541
CAS nummer: 63076-51-7