3-tert-Butoxypropionic acid 95+%
Leverantör: Apollo Scientific
Varning
OR7832-1G OR7832-5G
APOSOR7832-1GEA 3175 SEK
APOSOR7832-1G APOSOR7832-5G
3-tert-Butoxypropionic acid 95+%
3-tert-Butoxypropionic acid
Formel: C₇H₁₄O₃
MW: 146,19 g/mol
Kokpunkt: 70…72 °C/0,22mmHg
Densitet: 0,989 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD03426042
CAS nummer: 21150-73-2