5-Nitro-o-toluic acid 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Methyl-5-nitrobenzoic acid

OR7737-100G OR7737-10G
APOSOR7737-100GEA 1600 SEK
APOSOR7737-100G APOSOR7737-10G
5-Nitro-o-toluic acid 98%
5-Nitro-o-toluic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smältpunkt: 177…181 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007371
CAS nummer: 1975-52-6
EINECS: 217-829-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR