2-(4-Ethoxyphenyl)malondialdehyde 95+%
Leverantör: Apollo Scientific
OR7638-1G
OR7638-5G
APOSOR7638-1GEA
2175
SEK
APOSOR7638-1G
APOSOR7638-5G
2-(4-Ethoxyphenyl)malondialdehyde 95+%
2-(4-Ethoxyphenyl)malondialdehyde
Formel:
C₁₁H₁₂O₃ Smältpunkt: 128…130 °C Storage Temperature: Omgivande |
MDL Number:
MFCD03426048 CAS nummer: 849021-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...