3-Quinolineboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: Quinoline-3-boronic acid, Quinolin-3-ylboronic acid
OR7263-5G OR7263-25G
APOSOR7263-5GEA 175 SEK
APOSOR7263-5G APOSOR7263-25G
3-Quinolineboronic acid
3-Quinolineboronic acid
Formel: C₉H₈BNO₂
MW: 172,98 g/mol
Smältpunkt: 191…196 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02183527
CAS nummer: 191162-39-7