4-Propoxyphenylboronic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-n-Propoxybenzeneboronic acid
OR7256-100G OR7256-25G OR7256-5G
APOSOR7256-100GEA 9750 SEK
APOSOR7256-100G APOSOR7256-25G APOSOR7256-5G
4-Propoxyphenylboronic acid 97%
4-Propoxyphenylboronic acid
Formel: C₉H₁₃BO₃
MW: 180,01 g/mol
Smältpunkt: 104…110 °C
Storage Temperature: Omgivande
MDL Number: MFCD03427189
CAS nummer: 186497-67-6