6-Methoxy-2-naphthaleneboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid
OR7234-25G OR7234-5G OR7234-1G OR7234-10G
APOSOR7234-25GEA 1200 SEK
APOSOR7234-25G APOSOR7234-5G APOSOR7234-1G APOSOR7234-10G
6-Methoxy-2-naphthaleneboronic acid
6-Methoxy-2-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
MW: 202.02 g/mol
Smältpunkt: 311 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093087
CAS nummer: 156641-98-4