3,4,5-Trimethoxyaniline 98%

Leverantör: Apollo Scientific
Varning

OR7231-100G OR7231-25G OR7231-500G
APOSOR7231-100GEA 1025 SEK
APOSOR7231-100G APOSOR7231-25G APOSOR7231-500G
3,4,5-Trimethoxyaniline 98%
3,4,5-Trimethoxyaniline
Formel: C₉H₁₃NO₃
MW: 183,21 g/mol
Smältpunkt: 110…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008393
CAS nummer: 24313-88-0
EINECS: 246-154-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR