N,N-Dimethyl 2-boronobenzenesulphonamide
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(N,N-Dimethylsulphamoyl)benzeneboronic acid
OR7212-1G OR7212-5G
APOSOR7212-1GEA 2550 SEK
APOSOR7212-1G APOSOR7212-5G
N,N-Dimethyl 2-boronobenzenesulphonamide
N,N-Dimethyl 2-boronobenzenesulphonamide
Formel: C₈H₁₂BNO₄S
MW: 229,612 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD04039023
CAS nummer: 178432-25-2