3-Chloro-4-methylphenylboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid
OR7180-1G OR7180-5G OR7180-25G
APOSOR7180-1GEA 175 SEK
APOSOR7180-1G APOSOR7180-5G APOSOR7180-25G
3-Chloro-4-methylphenylboronic acid
3-Chloro-4-methylphenylboronic acid
Formel: C₇H₈BClO₂
MW: 170,40 g/mol
Smältpunkt: 228…232 °C
Storage Temperature: Omgivande
MDL Number: MFCD04039010
CAS nummer: 175883-63-3