4,4'-Biphenyldiboronic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: [1,1'-Biphenyl]-4,4'-diyldiboronic acid
OR7161-5G OR7161-10G
APOSOR7161-5GEA 1300 SEK
APOSOR7161-5G APOSOR7161-10G
4,4'-Biphenyldiboronic acid 97%
4,4'-Biphenyldiboronic acid
Formel: C₁₂H₁₂B₂O₄
MW: 241,85 g/mol
Smältpunkt: 300 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00151795
CAS nummer: 4151-80-8