2-Biphenylboronic acid 98%
Leverantör: Apollo Scientific
Varning
OR7160-25G OR7160-5G OR7160-100G
APOSOR7160-25GEA 895 SEK
APOSOR7160-25G APOSOR7160-5G APOSOR7160-100G
2-Biphenylboronic acid 98%
2-Biphenylboronic acid
Formel: C₁₂H₁₁BO₂
MW: 198,03 g/mol
Smältpunkt: 167…172 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00136929
CAS nummer: 4688-76-0