[Hydroxy(tosyloxy)iodo]benzene
Leverantör: Apollo Scientific
Varning
Synonyms: Koser Reagent, Phenyliodosohydroxy Tosylate
OR7131-100G OR7131-25G OR7131-500G
APOSOR7131-100GEA 960 SEK
APOSOR7131-100G APOSOR7131-25G APOSOR7131-500G
[Hydroxy(tosyloxy)iodo]benzene
[Hydroxy(tosyloxy)iodo]benzene
Formel: C₁₃H₁₃IO₄S
Smältpunkt: 131…137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011547
CAS nummer: 27126-76-7