4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Leverantör: Apollo Scientific
Fara
Synonyms: Pinacol Borane
OR7123-100G OR7123-25G OR7123-5G
APOSOR7123-100GEA 1725 SEK
APOSOR7123-100G APOSOR7123-25G APOSOR7123-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Reportedly couples with aryl halides to produce aryl boronates:J.Org.Chem.(1997)62,6458-6459.
Formel: C₆H₁₃BO₂
Kokpunkt: 42…43 °C/50mmHg
Densitet: 0,882 g/cm³ (25 °C)
Flampunkt: 5 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00674030
CAS nummer: 25015-63-8
UN: 3399
ADR: 4.3+3,II