Benzoimidazol-1-ylacetic acid 95%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-(1H-Benzo[d]imidazol-1-yl)acetic acid

OR7034-1G
APOSOR7034-1GEA 1200 SEK
APOSOR7034-1G
Benzoimidazol-1-ylacetic acid 95%
Benzoimidazol-1-ylacetic acid
Formel: C₉H₈N₂O₂
MW: 176.18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00956879
CAS nummer: 40332-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR