β-Bromostyrene (mixture of cis and trans isomers) 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (2-Bromovinyl)benzene, β-Bromostyrene
OR70303-100G OR70303-25G
APOSOR70303-100GEA 805 SEK
APOSOR70303-100G APOSOR70303-25G
β-Bromostyrene (mixture of cis and trans isomers) 98%
β-Bromostyrene (mixture of cis and trans isomers)
Formel: C₈H₇Br
MW: 183,05 g/mol
Kokpunkt: 110…112 °C/20,25mmHg
Smältpunkt: 7 °C
Densitet: 1,427 g/cm³ (25 °C)
Flampunkt: 102 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000185
CAS nummer: 103-64-0
EINECS: 203-131-3