5-Amino-2-methoxybenzoic acid
Leverantör: Apollo Scientific
|
Varning
|
OR70155-1G
OR70155-5G
APOSOR70155-1GEA
1400
SEK
APOSOR70155-1G
APOSOR70155-5G
5-Amino-2-methoxybenzoic acid
5-Amino-2-methoxybenzoic acid
Formel:
C₈H₉NO₃ MW: 167,16 g/mol Smältpunkt: 161…166 °C Storage Temperature: Omgivande |
MDL Number:
MFCD09260887 CAS nummer: 3403-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...