5-Bromobenzimidazole 98%

Leverantör: Apollo Scientific

OR70114-25G OR70114-5G
APOSOR70114-25GEA 930 SEK
APOSOR70114-25G APOSOR70114-5G
5-Bromobenzimidazole 98%
5-Bromobenzimidazole
Formel: C₇H₅BrN₂
Smältpunkt: 130…134 °C
Storage Temperature: Omgivande
MDL Number: MFCD00160001
CAS nummer: 4887-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR