(R)-1-(Pyridin-2-yl)ethanamine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (+)-2-[(1R)-1-Aminoethyl]pyridine
OR70108-10G OR70108-1G OR70108-5G
APOSOR70108-10GEA 8125 SEK
APOSOR70108-10G APOSOR70108-1G APOSOR70108-5G
(R)-1-(Pyridin-2-yl)ethanamine 98%
(R)-1-(Pyridin-2-yl)ethanamine
Formel: C₇H₁₀N₂
MW: 122,1677 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07776289
CAS nummer: 45695-03-2