(R)-2-Methylmorpholine hydrochloride
Leverantör: Apollo Scientific
Varning
OR70093-1G OR70093-250MG
APOSOR70093-1GEA 1375 SEK
APOSOR70093-1G APOSOR70093-250MG
(R)-2-Methylmorpholine hydrochloride
(R)-2-Methylmorpholine hydrochloride
Formel: C₅H₁₂ClNO
MW: 137,6100 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11506702
CAS nummer: 168038-14-0