2-Chloro-5-methylphenyl isocyanate, Technical Grade
Leverantör: Apollo Scientific
Fara
OR70050-100G OR70050-25G OR70050-5G
APOSOR70050-100GEA 3875 SEK
APOSOR70050-100G APOSOR70050-25G APOSOR70050-5G
2-Chloro-5-methylphenyl isocyanate, Technical Grade
2-Chloro-5-methylphenyl isocyanate
When melted, may contain small amount of sediment.
Formel: C₈H₆ClNO
MW: 167,59 g/mol
Smältpunkt: 39…42 °C
Flampunkt: 104 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01863693
CAS nummer: 40398-03-6