Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 1-Benzyl-3-ethoxycarbonyl-4-piperidone Hydrochloride, 1-Benzyl-4-oxo-3-piperidinecarboxylic acid ethyl ester hydrochloride
OR6944-100G OR6944-250G OR6944-25G
APOSOR6944-100GEA 2275 SEK
APOSOR6944-100G APOSOR6944-250G APOSOR6944-25G
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Formel: C₁₅H₁₉NO₃·HCl
Smältpunkt: 175 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012800
CAS nummer: 1454-53-1
EINECS: 215-929-9