3-Hydroxy-1-methylpiperidine 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1-Methylpiperidin-3-ol, N-Methyl-3-piperidinol
OR6933-1G
OR6933-25G
OR6933-5G
APOSOR6933-1GEA
175
SEK
APOSOR6933-1G
APOSOR6933-25G
APOSOR6933-5G
3-Hydroxy-1-methylpiperidine 98%
3-Hydroxy-1-methylpiperidine
Formel:
C₆H₁₃NO MW: 115,18 g/mol Kokpunkt: 76…78 °C/11mmHg Densitet: 0,999 g/cm³ (25 °C) Flampunkt: 70 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006495 CAS nummer: 3554-74-3 EINECS: 222-609-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...