Ethyl-1-benzyl-4-piperidinecarboxylate 96%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Benzylisonipecotic Acid Ethyl Ester, Ethyl 1-Benzyl-4-piperidinecarboxylate, 1-Benzyl-4-piperidinecarboxylic Acid Ethyl Ester, Ethyl 1-Benzylisonipecotate, Ethyl N-benzylpiperidine-4-carboxylate
OR6926-25G OR6926-5G OR6926-100G
APOSOR6926-25GEA 380 SEK
APOSOR6926-25G APOSOR6926-5G APOSOR6926-100G
Ethyl-1-benzyl-4-piperidinecarboxylate 96%
Ethyl-1-benzyl-4-piperidinecarboxylate
Formel: C₁₅H₂₁NO₂
MW: 247,34 g/mol
Kokpunkt: 122 °C/0,5mmHg
Storage Temperature: Omgivande
MDL Number: MFCD00040748
CAS nummer: 24228-40-8