(±)-Isonipecotic acid
Leverantör: Apollo Scientific
Varning
Synonyms: Piperidine-4-carboxylic acid, (±)-4-Piperidinecarboxylic acid, 4-Piperidinecarboxylic acid, Isonipecotic acid
OR6923-100G OR6923-1KG OR6923-25G
APOSOR6923-100GEA 410 SEK
APOSOR6923-100G APOSOR6923-1KG APOSOR6923-25G
(±)-Isonipecotic acid
(±)-Isonipecotic acid
Formel: C₆H₁₁NO₂
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006004
CAS nummer: 498-94-2
EINECS: 207-872-3