Ethyl-2-(5-bromo-2-thienyl)-2-oxoacetate 97%

Leverantör: Apollo Scientific
Varning

OR6743-10G OR6743-1G OR6743-5G
APOSOR6743-10GEA 5000 SEK
APOSOR6743-10G APOSOR6743-1G APOSOR6743-5G
Ethyl-2-(5-bromo-2-thienyl)-2-oxoacetate 97%
Ethyl-2-(5-bromo-2-thienyl)-2-oxoacetate
Formel: C₈H₇BrO₃S
Smältpunkt: 70…71 °C
Storage Temperature: Omgivande
CAS nummer: 22098-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR