6-Morpholin-4-ylpyridazine-3-carboxylic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 6-Morpholinopyridazine-3-carboxylic acid
OR6299-1G OR6299-250MG
APOSOR6299-1GEA 3950 SEK
APOSOR6299-1G APOSOR6299-250MG
6-Morpholin-4-ylpyridazine-3-carboxylic acid 97%
6-Morpholin-4-ylpyridazine-3-carboxylic acid
Formel: C₉H₁₁N₃O₃
MW: 209,21 g/mol
Smältpunkt: 203 °C
Storage Temperature: Omgivande
MDL Number: MFCD08235211
CAS nummer: 914637-36-8