N-Methyl-N-[(1-methyl-1H-pyrazol-3-yl)methyl]amine
Leverantör: Apollo Scientific
Synonyms: 1-Methyl-3-[(methylamino)methyl]-1H-pyrazole
OR6208-1G OR6208-50MG OR6208-250MG OR6208-500MG
APOSOR6208-1GEA 5675 SEK
APOSOR6208-1G APOSOR6208-50MG APOSOR6208-250MG APOSOR6208-500MG
N-Methyl-N-[(1-methyl-1H-pyrazol-3-yl)methyl]amine
N-Methyl-N-[(1-methyl-1H-pyrazol-3-yl)methyl]amine
Formel: C₆H₁₁N₃
MW: 125,17 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03419811
CAS nummer: 871825-57-9