2-Bromo-p-anisidine 96%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Bromo-4-methoxyaniline
OR61502-1G OR61502-5G OR61502-25G
APOSOR61502-1GEA 260 SEK
APOSOR61502-1G APOSOR61502-5G APOSOR61502-25G
2-Bromo-p-anisidine 96%
2-Bromo-p-anisidine
Formel: C₇H₈BrNO
MW: 202.0485 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09029057
CAS nummer: 32338-02-6
UN: 2811
ADR: 6.1 III