6-Cyano-2-naphthol
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
6-Cyano-2-naphthol, 6-Hydroxy-2-naphthonitrile
OR61500-25G
OR61500-5G
APOSOR61500-25GEA
2150
SEK
APOSOR61500-25G
APOSOR61500-5G
6-Cyano-2-naphthol
6-Cyano-2-naphthol
Formel:
C₁₁H₇NO MW: 169,1794 g/mol Smältpunkt: 165,5…170.5 °C Storage Temperature: Omgivande |
MDL Number:
MFCD01863480 CAS nummer: 52927-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...