5-Bromobenzoxazole

Leverantör: Apollo Scientific
Varning

Synonyms: 5-Bromobenzo[d]oxazole

OR61307-1G OR61307-5G OR61307-25G
APOSOR61307-1GEA 295 SEK
APOSOR61307-1G APOSOR61307-5G APOSOR61307-25G
5-Bromobenzoxazole
5-Bromobenzoxazole
Formel: C₇H₄BrNO
MW: 198,0200 g/mol
Smältpunkt: 37…40 °C
Storage Temperature: Omgivande
MDL Number: MFCD03095026
CAS nummer: 132244-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR