2-Aminopyrimidin-5-ol 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Amino-5-hydroxypyrimidine

OR61304-1G OR61304-5G
APOSOR61304-1GEA 4275 SEK
APOSOR61304-1G APOSOR61304-5G
2-Aminopyrimidin-5-ol 97%
2-Aminopyrimidin-5-ol
Formel: C₄H₅N₃O
MW: 111,1 g/mol
Smältpunkt: 198,6…199 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08062411
CAS nummer: 143489-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR