4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 4,5-Dimethoxy-2-nitrobenzyl chloroformate
OR61202-5G OR61202-1G
APOSOR61202-5GEA 4650 SEK
APOSOR61202-5G APOSOR61202-1G
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate
Formel: C₁₀H₁₀ClNO₆
MW: 275.65 g/mol
Smältpunkt: 120…125 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00143507
CAS nummer: 42855-00-5
UN: 3261
ADR: 8 II