Farnesol (mixture of isomers) 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Farnesol, 3,7,11-Trimethyldodeca-2,6,10-trien-1-ol, TRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL, 2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-
OR6039-25G OR6039-100G
APOSOR6039-25GEA 1125 SEK
APOSOR6039-25G APOSOR6039-100G
Farnesol (mixture of isomers) 95%
Farnesol (mixture of isomers)
Formel: C₁₅H₂₆O
MW: 222,37 g/mol
Kokpunkt: 149 °C/4mmHg
Densitet: 0,886 g/cm³ (25 °C)
Flampunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002918
CAS nummer: 4602-84-0
UN: 3082
ADR: 9+EHS,III