2-Methylisonicotinic acid, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Methylpyridine-4-carboxylic acid
OR60082-1G OR60082-250MG OR60082-5G
APOSOR60082-1GEA 260 SEK
APOSOR60082-1G APOSOR60082-250MG APOSOR60082-5G
2-Methylisonicotinic acid, Technical Grade
2-Methylisonicotinic acid
Formel: C₇H₇NO₂
MW: 137.14 g/mol
Smältpunkt: 295…300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00513435
CAS nummer: 4021-11-8