Thiophene-3-carboxylic acid amide
Leverantör: Apollo Scientific
Varning
Synonyms: Thiophene-3-carboxamide
OR60025-1G OR60025-5G
APOSOR60025-1GEA 1975 SEK
APOSOR60025-1G APOSOR60025-5G
Thiophene-3-carboxylic acid amide
Thiophene-3-carboxylic acid amide
Formel: C₅H₅NOS
MW: 127,17 g/mol
Smältpunkt: 181…184 °C
Storage Temperature: Kylskåp
MDL Number: MFCD05664206
CAS nummer: 51460-47-0