Ethyl glyoxylate 50% I toluen, Technical Grade
Leverantör: Apollo Scientific
Fara
OR59968-100G OR59968-500G OR59968-25G
APOSOR59968-100GEA 685 SEK
APOSOR59968-100G APOSOR59968-500G APOSOR59968-25G
Ethyl glyoxylate 50% I toluen, Technical Grade
Ethyl glyoxylate
Formel: C₄H₆O₃
MW: 102,09 g/mol
Kokpunkt: 110 °C
Densitet: 1,003 g/cm³ (25 °C)
Flampunkt: 7 °C
Storage Temperature: Omgivande
MDL Number: MFCD00044009
CAS nummer: 924-44-7
UN: 1294
ADR: 3,II