5-Chloro-2-nitrodiphenylamine 98%
Leverantör: Apollo Scientific
Synonyms:
5-Chloro-2-nitro-N-phenylaniline
OR59964-25G
OR59964-5G
APOSOR59964-25GEA
1300
SEK
APOSOR59964-25G
APOSOR59964-5G
5-Chloro-2-nitrodiphenylamine 98%
5-Chloro-2-nitrodiphenylamine
Formel:
C₁₂H₉ClN₂O₂ MW: 248,6700 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00007287 CAS nummer: 25781-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...