3-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%
Leverantör: Apollo Scientific
Varning
Synonyms: N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
OR59907-1G OR59907-5G
APOSOR59907-1GEA 1250 SEK
APOSOR59907-1G APOSOR59907-5G
3-(Dimethylcarbamoyl)phenylboronic acid pinacol ester 98%
3-(Dimethylcarbamoyl)phenylboronic acid pinacol ester
Formel: C₁₅H₂₂BNO₃
MW: 275.1500 g/mol
Smältpunkt: 90…94 °C
Storage Temperature: Omgivande
MDL Number: MFCD05863907
CAS nummer: 832114-07-5