2-Methylnicotinic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid
OR59824-1G OR59824-5G OR59824-25G
APOSOR59824-1GEA 175 SEK
APOSOR59824-1G APOSOR59824-5G APOSOR59824-25G
2-Methylnicotinic acid
2-Methylnicotinic acid
Formel: C₇H₇NO₂
MW: 137,14 g/mol
Smältpunkt: 228…230 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00013449
CAS nummer: 3222-56-8